EMAIL THIS PAGE TO A FRIEND
Hypertension (Dallas, Tex. : 1979)
Response to HYVET ambulatory blood pressure substudy.
PMID 23724417
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I1887
Indapamide, analytical standard, for drug analysis
C16H16ClN3O3S
P0094
Perindopril erbumine
C19H32N2O5 · C4H11N