EMAIL THIS PAGE TO A FRIEND
NASN school nurse (Print)
Epinephrine: the drug of choice for anaphylaxis.
PMID 23724601
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000883
Adrenaline with impurity F, European Pharmacopoeia (EP) Reference Standard
C9H13NO3