EMAIL THIS PAGE TO A FRIEND
The Journal of surgical research
Dobutamine-mediated heme oxygenase-1 induction via P13K and p38 MAPK inhibits high mobility group box 1 protein release and attenuates rat myocardial ischemia/reperfusion injury in vivo.
PMID 23726240