EMAIL THIS PAGE TO A FRIEND
Toxicology letters
TiO2, CeO2 and ZnO nanoparticles and modulation of the degranulation process in human neutrophils.
PMID 23726862
Abstract
Inflammation is frequently associated with nanoparticle (NP) exposures. Given that excessive polymorphonuclear neutrophil cell degranulation is a common feature of inflammatory disorders, and since these cells are key players in inflammation, we decided to test the hypothesis that NPs could act as modulators of degranulation in human neutrophils. TiO2, CeO2 and ZnO NPs slightly down-regulated cell surface expression of the granule marker CD35, but increased CD66b and CD63 expression, as assessed by flow cytometry. In addition, expression of myeloperoxidase, MMP-9 and albumin stored in azurophil, specific/gelatinase and secretrory granules, respectively, was significantly increased in the supernatants of NPs-induced neutrophils when compared to untreated cells. Moreover, NPs were more potent than the classical bacterial tripeptide N-formyl-methionine-leucine-phenylalanine (fMLP) agonist. Finally, TiO2 and CeO2 markedly increased the enzymatic activity of MMP-9 released into the supernatant, as assessed by gelatin zymography, while ZnO exerted only a modest effect. We conclude that NPs can differentially affect all steps involved during neutrophil degranulation, namely, cell surface expression of granule markers, liberation of proteins in the supernatants and enzymatic activity. These results are expected to be helpful to understand the toxicity of TiO2, CeO2 and ZnO.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
342955
Cerium(IV) oxide, fused, pieces, 3-6 mm, 99.9% trace metals basis
CeO2
544841
Cerium(IV) oxide, nanopowder, <25 nm particle size (BET)
CeO2
700290
Cerium(IV) oxide, nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis
CeO2
202975
Cerium(IV) oxide, powder, 99.995% trace metals basis
CeO2
211575
Cerium(IV) oxide, powder, <5 μm, 99.9% trace metals basis
CeO2
348341
Cerium(IV) oxide, powder, 90%
CeO2
22390
Cerium(IV) oxide, ≥99.0%
CeO2
289744
Cerium(IV) oxide, dispersion, 20 wt. % colloidal dispersion in 2.5% acetic acid, 30-50 nm avg. part. size
CeO2
643009
Cerium(IV) oxide, dispersion, nanoparticles, <25 nm particle size, 10 wt. % in H2O
CeO2
96479
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
OZn
14439
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
OZn
93632
Zinc oxide, analytical standard
OZn
204951
Zinc oxide, 99.999% trace metals basis
OZn
255750
Zinc oxide, 99.99% trace metals basis
OZn
205532
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
OZn
544906
Zinc oxide, nanopowder, <100 nm particle size
OZn
677450
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
OZn
752681
Zinc oxide, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.99% trace metals basis
OZn
721077
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
721085
Zinc oxide, dispersion, nanoparticles, 40 wt. % in ethanol, <130 nm particle size
721093
Zinc oxide, dispersion, nanoparticles, <110 nm particle size, 40 wt. % in butyl acetate