EMAIL THIS PAGE TO A FRIEND
International journal of orthodontics (Milwaukee, Wis.)
A new method to correct lingual rolling of lower molars (Amini twister).
PMID 23729138
Abstract
In this report, we present an uncomplicated approach to overcome lingual rolling of mandibular molars during orthodontic treatment, by using rectangular NiTi arch wire in twist fashion through the molar tube. The sequential steps of the twist manoeuvre on a typodont and a case report are exhibited in this article.