EMAIL THIS PAGE TO A FRIEND
Molecular immunology
Letter to the editor concerning the paper: Boltjes A, Op den Brouw ML, Biesta PJ, et al. Assessment of the effect of ribavirin on myeloid and plasmacytoid dendritic cells during interferon-based therapy of chronic hepatitis B patients [Molecular Immunology 53(1/2) (2013) 72-78].
PMID 23732513