EMAIL THIS PAGE TO A FRIEND
Occupational and environmental medicine
Obliterative bronchiolitis in workers laying up fiberglass-reinforced plastics with polyester resin and methylethyl ketone peroxide catalyst.
PMID 23739490