EMAIL THIS PAGE TO A FRIEND
Annals of the rheumatic diseases
Antibodies to modified citrullinated vimentin are associated with severe extra-articular manifestations in rheumatoid arthritis.
PMID 23740235