EMAIL THIS PAGE TO A FRIEND
Life sciences
Amelioration of cognitive, motor and endogenous defense functions with silymarin, piracetam and protocatechuic acid in the cerebral global ischemic rat model.
PMID 23743171
Abstract
The neuroprotective activities of silymarin, piracetam and protocatechuic acid ethyl ester (PCA) on cerebral global ischemic/reperfusion were evaluated in a rat model. A midline ventral incision was made in the throat region. The right and left common carotid arteries were located and a bilateral common carotid artery occlusion (BCCAO) was performed for 30min using atraumatic clamps followed by a 24h period of reperfusion. Neurological/behavioral functions (cognitive and motor), endogenous defense systems (lipid peroxidation, glutathione, catalase, and superoxide dismutase), reduced water content and infarct size and histopathological alterations were then studied. Silymarin and PCA treatments significantly improved cognitive, motor and endogenous defense functions, histopathological alterations, and, reduced both water content and infarct size compared to the vehicle-treated ischemic control group. Piracetam treatment improved neurological and histopathological alterations, reduced water content and infarct size, but failed to restore/prevent the impaired endogenous defense functions significantly. Silymarin showed better neuroprotection than piracetam and PCA in experimentally induced global ischemic/reperfusion and was able to facilitate mnemonic performance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E24859
Protocatechuic acid ethyl ester, 97%
C9H10O4