EMAIL THIS PAGE TO A FRIEND
Food science and technology international = Ciencia y tecnologia de los alimentos internacional
Relationship of dough thermomechanical properties with oil uptake, cooking and textural properties of instant fried noodles.
PMID 23744117
Abstract
Instant noodles were prepared from fifteen diverse wheat cultivars varying widely in their flour quality and dough rheology. Dough thermomechanical parameters obtained by Mixolab and flour analytical properties were correlated with the quality of instant noodles including oil uptake, cooking quality and textural attributes. The Mixolab parameters dough development time and dough stability showed significant positive correlation with cooking time, cooked weight, overall acceptability, hardness, springiness, cohesiveness and chewiness of noodles, while negatively correlated with oil uptake and cooking loss, therefore, exhibiting a marked positive effect on quality of instant noodles. Lower protein breakdown represented by C2 torque was also positively related with overall acceptability, hardness, springiness, cohesiveness and chewiness of noodles. Stickiness/adhesiveness of noodles was revealed to be mainly conferred by falling number values (R (2 )= 0.671) and damaged starch (R (2 )= 0.523) content of wheat flour samples. Flour samples with lesser values of protein content, sodium dodecyl sulphate sedimentation volume, thermal stability of proteins, dough stability and dough development time were found to be linked with poor noodle quality. Medium strong flours performed better in noodle making, while weaker flours demonstrated poor noodle quality. Dough rheology of good noodle making flours was characterized with higher dough development time, dough stability, C2, C3, C4 as well as C5 values. Noodles with higher overall acceptability showed a more continuous and uniform protein starch matrix in comparison to the poor counterparts.