EMAIL THIS PAGE TO A FRIEND
Revista de investigacion clinica; organo del Hospital de Enfermedades de la Nutricion
[Hyponatremia].
PMID 23745446
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F-005
Furosemide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C12H11ClN2O5S