EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Maternal and child undernutrition and overweight in low-income and middle-income countries.
PMID 23746772
Abstract
Maternal and child malnutrition in low-income and middle-income countries encompasses both undernutrition and a growing problem with overweight and obesity. Low body-mass index, indicative of maternal undernutrition, has declined somewhat in the past two decades but continues to be prevalent in Asia and Africa. Prevalence of maternal overweight has had a steady increase since 1980 and exceeds that of underweight in all regions. Prevalence of stunting of linear growth of children younger than 5 years has decreased during the past two decades, but is higher in south Asia and sub-Saharan Africa than elsewhere and globally affected at least 165 million children in 2011; wasting affected at least 52 million children. Deficiencies of vitamin A and zinc result in deaths; deficiencies of iodine and iron, together with stunting, can contribute to children not reaching their developmental potential. Maternal undernutrition contributes to fetal growth restriction, which increases the risk of neonatal deaths and, for survivors, of stunting by 2 years of age. Suboptimum breastfeeding results in an increased risk for mortality in the first 2 years of life. We estimate that undernutrition in the aggregate--including fetal growth restriction, stunting, wasting, and deficiencies of vitamin A and zinc along with suboptimum breastfeeding--is a cause of 3·1 million child deaths annually or 45% of all child deaths in 2011. Maternal overweight and obesity result in increased maternal morbidity and infant mortality. Childhood overweight is becoming an increasingly important contributor to adult obesity, diabetes, and non-communicable diseases. The high present and future disease burden caused by malnutrition in women of reproductive age, pregnancy, and children in the first 2 years of life should lead to interventions focused on these groups.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
207772
Iodine, ACS reagent, ≥99.8%, solid
I2
266426
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
I2
I3380
Iodine, ReagentPlus®, ≥99.8% (titration)
I2
376558
Iodine, flakes, ReagentPlus®, ≥99%
I2
57655
Iodine, puriss., ≥99.5% (RT), particles (round)
I2
03002
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
I2
31653
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Zn
96454
Zinc, purum, powder
Zn
209988
Zinc, dust, <10 μm, ≥98%
Zn
349410
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Zn
267619
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Zn
356018
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Zn
565148
Zinc, granular, 30-100 mesh, 99%
Zn
243469
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Zn
215503
Zinc, mossy, 99.99% trace metals basis
Zn
215481
Zinc, mossy, ≥99%
Zn
578002
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Zn
266353
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Zn
324930
Zinc, powder, <150 μm, 99.995% trace metals basis
Zn
267635
Zinc, shot, 5 mm, 99.999% trace metals basis
Zn
402583
Zinc, shot, <12 mm, 99.99% trace metals basis
Zn
266361
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Zn
267929
Zinc, wire, diam. 1.0 mm, 99.999% trace metals basis
Zn
716723
Zinc, shot, 99.9999%
Zn
749060
Zinc, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.995% trace metals basis
Zn
95653
Zinc (impurities), manufactured by MBH Analytical Ltd (41X Z2)
Zn
499552
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®
metal
Zn