EMAIL THIS PAGE TO A FRIEND
Cell
Reshaping antibody diversity.
PMID 23746848
Abstract
Some species mount a robust antibody response despite having limited genome-encoded combinatorial diversity potential. Cows are unusual in having exceptionally long CDR H3 loops and few V regions, but the mechanism for creating diversity is not understood. Deep sequencing reveals that ultralong CDR H3s contain a remarkable complexity of cysteines, suggesting that disulfide-bonded minidomains may arise during repertoire development. Indeed, crystal structures of two cow antibodies reveal that these CDR H3s form a very unusual architecture composed of a β strand "stalk" that supports a structurally diverse, disulfide-bonded "knob" domain. Diversity arises from somatic hypermutation of an ultralong DH with a severe codon bias toward mutation to cysteine. These unusual antibodies can be elicited to recognize defined antigens through the knob domain. Thus, the bovine immune system produces an antibody repertoire composed of ultralong CDR H3s that fold into a diversity of minidomains generated through combinations of somatically generated disulfides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3290000
Cysteine hydrochloride monohydrate, European Pharmacopoeia (EP) Reference Standard
C3H7NO2S · HCl · H2O
14495 L-Cysteine 0.1 M solution
C7880
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
C3H7NO2S · HCl · H2O
C6852
L-Cysteine hydrochloride monohydrate, from non-animal source, cell culture tested, meets EP, USP testing specifications
C3H7NO2S · HCl · H2O
C5610
L-Cysteine hydrochloride monohydrate, PharmaGrade, Ajinomoto, EP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C3H7NO2S · HCl · H2O
30122
L-Cysteine hydrochloride monohydrate, tested according to Ph.Eur.
C3H7NO2S · HCl · H2O
W779989
L-Cysteine hydrochloride monohydrate, FG, produced by Wacker Chemie AG, Burghausen, Germany, Food, 20 mesh
C3H7NO2S · HCl · H2O
W779865
L-Cysteine hydrochloride monohydrate, 99%, FG, produced by Wacker Chemie AG, Burghausen, Germany
C3H7NO2S · HCl · H2O
PHR1102
L-Cysteine hydrochloride monohydrate, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO2S · HCl · H2O
30129
L-Cysteine hydrochloride monohydrate, BioUltra, ≥99.0% (RT)
C3H7NO2S · HCl · H2O
30130
L-Cysteine hydrochloride monohydrate, ≥99.0% (RT)
C3H7NO2S · HCl · H2O
778451
L-Cysteine hydrochloride monohydrate, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, 98.5-101.0% (Ph.Eur (2.2.20))
C3H7NO2S · HCl · H2O