EMAIL THIS PAGE TO A FRIEND
Phytomedicine : international journal of phytotherapy and phytopharmacology
Protective effect of homonojirimycin from Commelina communis (dayflower) on influenza virus infection in mice.
PMID 23746956
Abstract
The effects of homonojirimycin (HNJ), one of alkaloids from Commelina communis L., on protection against influenza virus infection in mice were investigated. HNJ was found to improve the survival rate, prolong the mean survival time and reduce virus yields in lungs on days 4 and 6 post-infection (p.i.), after the agent had been orally administered to the mice from 2 days before infection to 6 days p.i. Administration of HNJ (1 mg/kg) significantly increased interferon (IFN)-γ and interleukin (IL)-10 levels but decreased tumor necrosis factor (TNF)-α and IL-6 levels in serum and lungs of influenza-infected mice on days 2, 4 or 6 p.i. These results showed that HNJ exerted protection against influenza virus infection and produced effective immune responses in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
08012
1-Deoxynojirimycin, analytical standard
C6H13NO4