EMAIL THIS PAGE TO A FRIEND
Recenti progressi in medicina
[Prevention of post-operative recurrence in Crohn's disease: a critical review of randomized controlled trials and meta-analyses of different therapeutic strategies].
PMID 23748641
Abstract
Surgery is an almost inevitable event in Crohn's disease but is not curative; post-operative recurrence follows a sequential and predictable course. Prevention of post-operative recurrence in Crohn's disease is therefore a relevant problem in the management of the disease. Several drugs have been evaluated to decrease the risk of recurrence: these include mesalazine, antibiotics, probiotics, budesonide, thiopurines and biologic agents. This review focuses on the randomised controlled trials and meta-analyses addressing different drugs and strategies for preventing post-operative recurrence in Crohn's disease.