EMAIL THIS PAGE TO A FRIEND
Marine biotechnology (New York, N.Y.)
Isolation of fucoxanthin and highly unsaturated monogalactosyldiacylglycerol from brown alga Fucus evanescens C Agardh and in vitro investigation of their antitumor activity.
PMID 23748883
Abstract
Fucoxanthin (FX) and highly unsaturated monogalactosyldiacylglycerol (MGDG) were isolated from the ethanol extract of brown alga Fucus evanescens. Their structures were identified by nuclear magnetic resonance, complemented by electrospray ionization mass spectrometry (ESIMS). MGDG was identified as 1-O-(5Z,8Z,11Z,14Z,17Z-eicosapentanoyl)-2-O-(6Z,9Z,12Z,15Z-octadecatetraenoyl)-3-O-β-D-galactopiranosyl-sn-glycerol. Antitumor activity of these compounds was tested on human melanoma (SK-MEL-28) cells. MGDG and FX inhibited the growth of human melanoma cells in a dose-dependent manner. IC50 values for growth inhibition were 104 and 114 μM, correspondently.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6932
Fucoxanthin, carotenoid antioxidant
C42H58O6