EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
The effects of cyclooxygenase and nitric oxide synthase inhibition on oxidative stress in isolated rat heart.
PMID 23749198
Abstract
Despite the widespread clinical use of cyclooxygenase (COX) inhibitors, dilemmas still exist about potential impact of these drugs on cardiovascular system. The present study was aimed to estimate the effects of different COX inhibitors (meloxicam, acetylsalicylic acid [ASA], and SC-560) on oxidative stress in isolated rat heart, with special focus on L-arginine/NO system. The hearts of male Wistar albino rats (total number n = 96, each group 12 rats, 8 weeks old, body mass 180-200 g) were retrogradely perfused according to the Langendorff technique at gradually increased perfusion pressure (40-120 cmH2O). After control experiments the hearts were perfused with the following drugs: 100 μmol/l ASA (Aspirin), alone or in combination with 30 μmol/l L-NAME, 0.3 μmol/l meloxicam (movalis) with or without 30 μmol/l L-NAME, 3 μmol/l meloxicam (alone or in combination with 30 μmol/l L-NAME), 30 μmol/l L-NAME, and administration of 0.25 μmol/l SC-560. In samples of coronary venous effluent the following oxidative stress markers were measured spectrophotometrically: index of lipid peroxidation (measured as thiobarbituric acid reactive substances [TBARS]), superoxide anion radical release (O2(-)), and hydrogen peroxide (H2O2). While ASA was found to have an adverse influence on redox balance in coronary circulation, and coronary perfusion, meloxicam and SC-560 do not negatively affect the intact model of the heart. Furthermore, all effects were modulated by NOS inhibition. It seems that interaction between COX and L-arginine/NO system truly exists in coronary circulation, and can be one of the possible causes for achieved effects. That means: those effects induced by different inhibitors of COX are modulated by subsequent inhibition of NOS.