EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
HPLC column-switching technique for sample preparation and fluorescence determination of propranolol in urine using fused-core columns in both dimensions.
PMID 23754331
Abstract
A new and fast high-performance liquid chromatography (HPLC) column-switching method using fused-core columns in both dimensions for sample preconcentration and determination of propranolol in human urine has been developed. On-line sample pretreatment and propranolol preconcentration were performed on an Ascentis Express RP-C-18 guard column (5 × 4.6 mm), particle size, 2.7 μm, with mobile phase acetonitrile/water (5:95, v/v) at a flow rate of 2.0 mL min(-1) and at a temperature of 50 °C. Valve switch from pretreatment column to analytical column was set at 4.0 min in a back-flush mode. Separation of propranolol from other endogenous urine compounds was achieved on the fused-core column Ascentis Express RP-Amide (100 × 4.6 mm), particle size, 2.7 μm, with mobile phase acetonitrile/water solution of 0.5% triethylamine, pH adjusted to 4.5 by means of glacial acetic acid (25:75, v/v), at a flow rate of 1.0 mL min(-1) and at a temperature of 50 °C. Fluorescence excitation/emission detection wavelengths were set at 229/338 nm. A volume of 1,500 μL of filtered urine sample solution was injected directly into the column-switching HPLC system. The total analysis time including on-line sample pretreatment was less than 8 min. The experimentally determined limit of detection of the method was found to be 0.015 ng mL(-1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
581311-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 1 mm
581364-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 1 mm
581312-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 2 cm × 2.1 mm
581300-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 2.1 mm
581301-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 2.1 mm
581302-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 2.1 mm
581306-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 3 cm × 3 mm
581307-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 3 mm
581308-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 3 mm
581316-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 3 cm × 4.6 mm
581320-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 4.6 mm
581321-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 4.6 mm
581322-U Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
581327-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 3 cm × 2.1 mm
581303-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
581326-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
581304-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
581305-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
581328-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 2 cm × 3 mm
581369-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 3 cm × 3 mm
581323-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
581324-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
581325-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
581342-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 10 mm
581343-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
581344-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 21.2 mm
581346-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 21.2 mm
581347-U Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
581351-U Ascentis® C18 HPLC Column, 10 μm particle size, L × I.D. 25 cm × 4.6 mm
581352-U Ascentis® C18 HPLC Column, 10 μm particle size, L × I.D. 5 cm × 10 mm