EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
China's skies: a complex recipe for pollution with no quick fix.
PMID 23755386
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295698
Sulfur dioxide, ≥99.9%
O2S