EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
Replay: being sensitive: to specify when amino acid tracers accumulate in a brain lesion.
PMID 23757488