EMAIL THIS PAGE TO A FRIEND
Journal of Orthopaedic Surgery and Research
Strontium enhances osseointegration of calcium phosphate cement: a histomorphometric pilot study in ovariectomized rats.
PMID 23758869
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3786
Potassium phosphate dibasic, ACS reagent, ≥98%
HK2O4P