EMAIL THIS PAGE TO A FRIEND
Nederlands tijdschrift voor geneeskunde
[Hallucinogen persisting perception disorder after ecstasy use].
PMID 23759176
Abstract
Hallucinogen persisting perception disorder (HPPD) is a relatively unknown complication of ecstasy use. As a result, the diagnosis is often missed. The prevalence of HPPD is probably low. Few patients seek medical help; HPPD is however widely discussed by drug users on internet fora. We describe a 23-year-old male with HPPD after ecstasy use. Besides visual complaints, including seeing little spots ('visual snow'), the patient suffered from anxiety and depressive symptoms. We treated him with citalopram with good results. In patients with HPPD combined with anxiety or depressive complaints, citalopram could be a good treatment option. Further research into treatment is necessary.
Related Materials