EMAIL THIS PAGE TO A FRIEND
Experimental biology and medicine (Maywood, N.J.)
Rutin suppresses atopic dermatitis and allergic contact dermatitis.
PMID 23760007
Abstract
Atopic dermatitis (AD) and allergic contact dermatitis (ACD) is a common allergic inflammatory skin disease caused by a combination of eczematous, scratching, pruritus and cutaneous sensitization with allergens. The aim of our study was to examine whether rutin, a predominant flavonoid having anti-inflammatory and antioxidative potential, modulates AD and ACD symptoms. We established an atopic dermatitis model in BALB/c mice by repeated local exposure of house dust mite (Dermatophagoides farinae) extract (DFE) and 2,4-dinitrochlorobenzene (DNCB) to the ears. In addition, 2,4-dinitroflourobenzene-sensitized a local lymph node assay was used for the ACD model. Repeated alternative treatment of DFE/DNCB caused AD symptoms. Topical application of rutin reduced AD based on ear thickness and histopathological analysis, in addition to serum IgE levels. Rutin inhibited mast cell infiltration into the ear and serum histamine level. Rutin suppressed DFE/DNCB-induced expression of interleukin (IL)-4, IL-5, IL-13, IL-31, IL-32 and interferon (INF)-γ in the tissue. In addition, rutin suppressed ACD based on ear thickness and lymphocyte proliferation, serum IgG2a levels, and expression of INF-γ, IL-4, IL-5, IL-10, IL-17 and tumour necrosis factor-α in ACD ears. This study demonstrates that rutin inhibits AD and ACD, suggesting that rutin might be a candidate for the treatment of allergic skin diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73177
1-Fluoro-2,4-dinitrobenzene, for HPLC derivatization, LiChropur, ≥99.0% (GC)
C6H3FN2O4