EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and chemistry
Silver nanoparticle toxicity to Daphnia magna is a function of dissolved silver concentration.
PMID 23761010
Abstract
The most persistent question regarding the toxicity of silver nanoparticles (AgNPs) is whether this toxicity is due to the nanoparticles themselves or the silver ions (Ag(+)) they release. The present study investigates the role of surface coating and the presence of dissolved organic carbon on the toxicity of AgNPs to Daphnia magna and tests the hypothesis that the acute toxicity of AgNPs is a function of dissolved Ag produced by nanoparticle dissolution. Toxicity of silver nitrate (AgNO3) and AgNPs with surface coatings-gum arabic (AgGA), polyethylene glycol (AgPEG), and polyvinylpyrrolidone (AgPVP)-at 48 h was assessed in US Environmental Protection Agency moderately hard reconstituted water alone and augmented with Suwannee River dissolved organic carbon (DOC). As expected, AgNO3 was the most toxic to D. magna and AgPVPs were the least toxic. In general, Suwannee River DOC presence reduced the toxicity of AgNO3, AgGAs, and AgPEG, while the toxicity of AgPVPs was unaffected. The measured dissolved Ag concentrations for all AgNPs and AgNO3 at the 48-h median lethal concentration in moderately hard reconstituted water were similar. The presence of Suwannee River DOC decreased the ratio of measured dissolved Ag to measured total Ag concentration. These results support the hypothesis that toxicity of AgNPs to D. magna is a function of dissolved Ag concentration from these particles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
02286
Kollidon® 25
77627
Poly(vinylpolypyrrolidone), ~110 μm particle size
PVP10
Polyvinylpyrrolidone, average mol wt 10,000
81420
Polyvinylpyrrolidone, K 30
PVP360
Polyvinylpyrrolidone, average mol wt 360,000
437190
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
856568
Polyvinylpyrrolidone, powder, average Mw ~55,000
81440
Polyvinylpyrrolidone, K 90
234257
Polyvinylpyrrolidone, powder, average Mw ~29,000
P5288
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
P0930
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
P2307
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
90268
Polyvinylpyrrolidone, K 25, tested according to Ph Eur
81430
Polyvinylpyrrolidone solution, K 60, 45% in H2O
PHR1250
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
P2472
Povidone, meets USP testing specifications
P2660000
Povidone, European Pharmacopoeia (EP) Reference Standard
209139
Silver nitrate, ACS reagent, ≥99.0%
AgNO3
S6506
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
AgNO3
10220
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
AgNO3
31630
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
AgNO3
85228
Silver nitrate, puriss. p.a., ≥99.5% (AT)
AgNO3
85229
Silver nitrate, tested according to Ph Eur
AgNO3
11691
Silver standard for ICP, for ICP, ready-to-use, in nitric acid
AgNO3