EMAIL THIS PAGE TO A FRIEND
Biomedical optics express
Simple buffers for 3D STORM microscopy.
PMID 23761850
Abstract
3D STORM is one of the leading methods for super-resolution imaging, with resolution down to 10 nm in the lateral direction, and 30-50 nm in the axial direction. However, there is one important requirement to perform this type of imaging: making dye molecules blink. This usually relies on the utilization of complex buffers, containing different chemicals and sensitive enzymatic systems, limiting the reproducibility of the method. We report here that the commercial mounting medium Vectashield can be used for STORM of Alexa-647, and yields images comparable or superior to those obtained with more complex buffers, especially for 3D imaging. We expect that this advance will promote the versatile utilization of 3D STORM by removing one of its entry barriers, as well as provide a more reproducible way to compare optical setups and data processing algorithms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92544 Abberior® FLIP 565, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
79189 Abberior® FLIP 565, NHS ester, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
HPA039408
Anti-CEP152 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
50283 Anti-Mouse IgG - Atto 655 antibody produced in goat, ~1 mg/mL IgG
78519 Anti-Rabbit IgG - Atto 655 antibody produced in goat, ~1 mg/mL IgG
93711 Atto 655, BioReagent, suitable for fluorescence, ≥85% (HPLC)
92009 Atto 655 alkyne, BioReagent, suitable for fluorescence
14918 Atto 655 amine, suitable for fluorescence
11774 Atto 655 azide, BioReagent, suitable for fluorescence
73948 Atto 655 iodoacetamide, suitable for fluorescence
73919 Atto 655 Protein Labeling Kit
06966 Atto 655-Biotin, BioReagent, suitable for fluorescence, ≥95.0% (HPCE)
80661 Atto 655-maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling rate)
02744 Atto 655-Streptavidin, BioReagent, suitable for fluorescence
SAB5200570
Monoclonal Anti-Adam22 - Atto 655 antibody produced in mouse, clone S46-30, purified immunoglobulin
SAB5200588
Monoclonal Anti-Adam22 - Atto 655 antibody produced in mouse, clone S57-2, purified immunoglobulin
SAB5200138
Monoclonal Anti-Alginate - Atto 655 antibody produced in mouse, clone 4B10-1C5, purified immunoglobulin
SAB5200156
Monoclonal Anti-Alginate - Atto 655 antibody produced in mouse, clone 3G4-1F5, purified immunoglobulin
SAB5200966
Monoclonal Anti-Alpha - Atto 655 antibody produced in mouse, clone S330A-51, purified immunoglobulin
SAB5201002
Monoclonal Anti-Alpha 2C Adrenergic Receptor - Atto 655 antibody produced in mouse, clone S330A-80, purified immunoglobulin
SAB5201056
Monoclonal Anti-Amigo-1 - Atto 655 antibody produced in mouse, clone S86-36, purified immunoglobulin
SAB5200372
Monoclonal Anti-Ankyrin B - Atto 655 antibody produced in mouse, clone S105-13, purified immunoglobulin
SAB5200444
Monoclonal Anti-Ankyrin G - Atto 655 antibody produced in mouse, clone S106-20, purified immunoglobulin
SAB5200624
Monoclonal Anti-Ankyrin-B - Atto 655 antibody produced in mouse, clone S105-17, purified immunoglobulin
SAB5200858
Monoclonal Anti-Asic1 - Atto 655 antibody produced in mouse, clone S271-44, purified immunoglobulin
SAB5201362
Monoclonal Anti-Ataxin - Atto 655 antibody produced in mouse, clone S76-8, purified immunoglobulin
SAB5201686
Monoclonal Anti-Ataxin 1 - Atto 655 antibody produced in mouse, clone S65-37, purified immunoglobulin
SAB5201218
Monoclonal Anti-Atf4 - Atto 655 antibody produced in mouse, clone S360A-24, purified immunoglobulin
SAB5200876
Monoclonal Anti-Brevican - Atto 655 antibody produced in mouse, clone S294A-6, purified immunoglobulin
SAB5200462
Monoclonal Anti-Cav3.1 - Atto 655 antibody produced in mouse, clone S178A-9, purified immunoglobulin