EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[Effect of hemantane on bioelectric activity of brain in mice with MPTP-induced Parkinson syndrome].
PMID 23762980
Abstract
A single intraperitoneal injection of MPTP neurotoxin (30 mg/kg) in C57BL/6 mice causes desynchronization of EEG with a decrease of theta-1 activity and a growth of beta activity in the interval of 15-30 Hz. Subchronic administration of the new antiparkinsonian drug hemantane (injection form) in a dose of 10 mg/kg makes the power of MPTP-induced beta oscillations less pronounced and leads to its reliable decrease within 24 h. This effect of hemantane administration was manifested during the entire period of observations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
100277
Adamantane, ≥99%
C10H16