EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[Psychomodulating activity of phenotropil in experimental hyperthyroidism].
PMID 23762984
Abstract
Psychoimmunomodulating action of phenotropil has been shown on the model of experimental hyperthyroidism. It has been found that the drug (14-days i.p. injection in a dose of 25 mg/kg) is capable of restoring the cellular and humoral immunoreactivity and improve psychoemotonal state of animals by eliminating disturbances in the behavior reactions that appear as a result of the induced hyperthyroidism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5295
Piracetam
C6H10N2O2