EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[Effect of Tri-V-Plus preparation on experimental echinococcal pathology in rats].
PMID 23762988
Abstract
The effects of Tri-V-Plus preparation in experimental rats with echinococcal pathology have been studied by monitoring the indices of free-radical oxidation of lipids, oxidative degradation of proteins, and variation of the content of nitric oxide during experimental echinococcosis and upon the introduction of Tri-V-Plus as a therapeutic agent.