EMAIL THIS PAGE TO A FRIEND
EMS world
How can tranexamic acid help in EMS? TXA offers new hope for an old challenge in traumatic hemorrhagic shock.
PMID 23763065
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1810000
Tranexamic acid, European Pharmacopoeia (EP) Reference Standard
C8H15NO2