EMAIL THIS PAGE TO A FRIEND
Anesthesia progress
New FDA black box warning for codeine: how will this affect dentists?
PMID 23763556
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5901
Codeine monohydrate, analytical standard
C18H21NO3 · H2O