EMAIL THIS PAGE TO A FRIEND
Anesthesia progress
Prevention of sevoflurane delirium and agitation with propofol.
PMID 23763562
Abstract
Emergence delirium and agitation (EAD) associated with sevoflurane general anesthesia are very commonly observed in young children. Such events pose a risk for injury as well as decreased parental satisfaction, especially in the ambulatory and office-based setting. This article reviews the different approaches described in the literature to reduce EAD. A novel approach using a Bispectral Index System (BIS)-guided anesthesia with propofol washout technique is proposed as a viable and effective approach to prevent EAD.
Related Materials