EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Separation of the enantiomeric intermediates of some platelet-activating factor analogues on a naphthylalanine-type Pirkle column.
PMID 2376609
Abstract
A chiral high-performance liquid-chromatographic separation method was developed for the analysis of some glycerol-based, ether-linked platelet-activating factor precursors. Using a D-naphthylalanine-type Pirkle-column and tetrahydrofuran-hexane eluents, the retention-structure relationships were determined for a large number of glycerol ether derivatives. The derivatives leading to the highest chiral selectivity factors in the analytical separations were surveyed for their applicability in large-scale isolation of chiral synthetic intermediates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5637
3-(1-Naphthyl)-DL-alanine
C13H13NO2