EMAIL THIS PAGE TO A FRIEND
Folia biologica
The content of conjugated linoleic acid (CLA) isomer groups in milk of two Polish sheep breeds determined by silver ion liquid chromatography (Ag(+)-HPLC).
PMID 23767301
Abstract
Sheep milk is rich in CLA isomers which are biologically active components influencing human health. There are four geometric CLA isomer pairs: cis, trans; trans, cis; trans, trans and cis, cis. The aim of the present study was the analysis of CLA isomer groups content by Ag+-HPLC in milk fat of Zelazneńska (ZS) and Wrzosówka (WS) sheep breeds. The ewes of both breeds were kept under the same environmental and nutritional conditions. Milk samples were collected from 60 suckling ewes (30 from each breed), at the age of 3-4 years and in their 4th week of lactation. A higher total amount of all CLA isomer groups was obtained in milk of ZS ewes, however, this result was statistically insignificant. The percentage of the main c9, t11 isomer in total CLA was higher in Wrzosówka milk (68% vs. 74%). The content of the trans, trans isomer group in milk fat of the studied breeds was similar. The percentage of this group in total CLA in milk of WS and ZS constitutes 7.2% and 7.7%, respectively. The amount of cis,cis isomers in milk fat of ZS was higher thanWS (P < or = 0.01).