EMAIL THIS PAGE TO A FRIEND
Journal of dermatological science
A novel mutation, c.699C>G (p.C233W), in the LIPH gene leads to a loss of the hydrolytic activity and the LPA6 activation ability of PA-PLA1α in autosomal recessive wooly hair/hypotrichosis.
PMID 23768866
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L3295 Phospholipase A1 from Aspergillus oryzae