EMAIL THIS PAGE TO A FRIEND
Ecotoxicology and environmental safety
Pseudo-basal levels of and distribution of anti-oxidant enzyme biomarkers in Eisenia fetida and effect of exposure to phenanthrene.
PMID 23769124
Abstract
In the paper, the pseudo-basal levels of anti-oxidant system in different earthworm life stages (juvenile and adult) and the pseudo-basal distribution in different regions of adult earthworms (pre-clitellum, clitellum and post-clitellum) were studied using filter contact tests. The effects of phenanthrene (PHE) at different exposure levels on anti-oxidant enzymes along the earthworm body were also investigated after 24 and 48h of exposure. The pseudo-basal levels of the anti-oxidant enzymes varied during the different growth phase, and results indicated that earthworm has a low oxidative risk and SOD plays important roles during the development whereas CAT and POD are more important in maintain the low ROS level in adult earthworm. The pseudo-basal distribution of the anti-oxidant enzymes along the earthworms was heterogeneous and MDA mainly located in clitellum. POD in pre-clitellum, SOD in clitellum and CAT in post-clitellum were important to eliminate excess total ROS. Time of exposure impacted the anti-oxidant enzyme activities and their distribution patterns along earthworms, from the viewpoint of which supported that exposure time was an environment stress factors. In a short exposure time (24 h), CAT and SOD in the three regions, POD in pre-clitellum and clitellum might be good indicator to a low PHE stress level (0.0629 μg cm(-2) treatments). In a long exposure time (48 h), only SOD in clitellum is a good indicator to both low and high PHE stress (0.629 μg cm(-2) treatments). Earthworm biomembrane system inflicted no oxidative damage until the stress magnitude reached or exceeds the level of exposure in low PHE concentration condition for 48 h.
Related Materials