EMAIL THIS PAGE TO A FRIEND
The Lancet. Neurology
Loa loa: neglected neurology and nematodes.
PMID 23769594
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I8898
Ivermectin