EMAIL THIS PAGE TO A FRIEND
Human immunology
Interleukin-6 promoter polymorphisms -174 G/C in Brazilian patients with systemic lupus erythematosus.
PMID 23770252
Abstract
To investigate the association of the IL-6 gene promoter polymorphism (-174 G/C) with SLE susceptibility and disease features in Brazilian patients, a case-control study of 80 lupus cases and 60 volunteer healthy women was performed. Genotyping was carried out by polymerase chain reaction and PCR product was digested by HSP92II restriction enzyme, being after visualized in polyacrylamide gel. There were significant differences in the distribution of the IL-6 gene C/C polymorphism between the SLE and control groups (χ(2) = 8.668; P = 0.0032). Individual carriers of the variant allele G had a 1.98 (95% CI: 1.0844-3.6353)-fold increased risk for SLE. Besides, association was observed in frequency of C allele (P = 0.0247; OR = 0.5036) between SLE patients and control groups. This study presents preliminary evidence for association between IL-6 polymorphism and SLE susceptibility in a Northeast population from Brazil.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R6142 Acc I from Acinetobacter calcoaceticus, Restriction Enzyme
R6885 Alu I from Arthrobacter luteus, Restriction Enzyme
R8631
Bcl I from Bacillus caldolyticus, Restriction Enzyme
R6377
Bgl II from Bacillus licheniformis, Restriction Enzyme
R3131
Bln I from Brevibacterium linens, buffered aqueous glycerol solution
R3635
Bsm I from Bacillus stearothermophilus NUB 36, Restriction Enzyme
R4253 BstE II from Bacillus stearothermophilus ET, Restriction Enzyme
R1761
Cfo I from Clostridium formicoaceticum, Restriction Enzyme
R7763
Cla I from Caryophanon latum L, Restriction Enzyme
R4256
Dde I from Desulfovibrio desulfuricans strain Norway, Restriction Enzyme
R4381
Dra I from Deinococcus radiophilus, Restriction Enzyme
R3884 EclX I from Enterobacter cloacae 590, Restriction Enzyme
R6265
EcoR I from Escherichia coli BS5, Restriction Enzyme
R2756
EcoR V from Escherichia coli, buffered aqueous glycerol solution
R5628 Hae III from Haemophilus aegyptius, Restriction Enzyme
R1632
Mva I from Micrococcus varians Rfl 19, Restriction Enzyme
R8761 Nco I from Nocardia corallina, Restriction Enzyme
R5634
Nhe I from Neisseria mucosa heidelbergensis, Restriction Enzyme
R8506
Not I from Nocardia otidiscaviarum, Restriction Enzyme
R5884 Nsi I from Neisseria sicca, Restriction Enzyme
R7023
Pst I from Providencia stuartii, Restriction Enzyme
R1508
Pvu I from Proteus vulgaris, Restriction Enzyme
R2631
Pvu II from Proteus vulgaris, Restriction Enzyme
R4756 Rsa I from Rhodopseudomonas sphaeroides, Restriction Enzyme
R0754 Sal I from Streptomyces albus G, Restriction Enzyme
R8256
Sfi I from Streptomyces fimbriatus, Restriction Enzyme
R7135 Sph I from Streptomyces phaeochromogenes, Restriction Enzyme
R7260
Xba I from Xanthomonas badrii, Restriction Enzyme
R6379
Xho I from Xanthomonas holcicola, Restriction Enzyme