EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Interaction of Schiff base ligand with tin dioxide nanoparticles: optical studies.
PMID 23770505
Abstract
Interaction between 1,4 Bis ((2-Methyl) thio) Phenylamino methyl benzene (BMTPMB) Schiff base with tin dioxide nanoparticles (SnO2 NPs) of various concentrations in methanol have been studied using UV-Visible and Fluorescence spectroscopic techniques. The low value of Stern-Volmer quenching constant and non-linear plot of Benesi-Hildebrand equation suggests the less affinity of SnO2 NPs towards the adsorption of BMTPMB Schiff base. The Scott equation has been employed to determine molar absorptivity of the Schiff base-NPs system.