EMAIL THIS PAGE TO A FRIEND
The journal of evidence-based dental practice
Inferior alveolar nerve block with articaine supplemented with articaine buccal infiltration gives better success rate than lidocaine, during endodontic management of patients with symptomatic irreversible pulpitis.
PMID 23773469
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000079
Articaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C13H20N2O3S · HCl