EMAIL THIS PAGE TO A FRIEND
BMJ (Clinical research ed.)
Europe recommends ban on common plasma substitute.
PMID 23775859
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6382
Hydroxyethyl starch