EMAIL THIS PAGE TO A FRIEND
Nihon Naika Gakkai zasshi. The Journal of the Japanese Society of Internal Medicine
[Case report ; a case of Bardet-Biedl syndrome with a pituitary tumor and growth hormone deficiency].
PMID 23777161
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8648 Somatotropin (STH) from porcine pituitary