EMAIL THIS PAGE TO A FRIEND
BMJ (Clinical research ed.)
Inhaled adrenaline no better than inhaled saline for babies with bronchiolitis.
PMID 23778881
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000883
Adrenaline with impurity F, European Pharmacopoeia (EP) Reference Standard
C9H13NO3