EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Susceptibility breakpoint for enrofloxacin against swine Salmonella spp.
PMID 23784134
Abstract
Susceptibility breakpoints are crucial for prudent use of antimicrobials. This study has developed the first susceptibility breakpoint (MIC ≤ 0.25 μg/ml) for enrofloxacin against swine Salmonella spp. based on wild-type cutoff (COWT) and pharmacokinetic-pharmacodynamic (PK-PD) cutoff (COPD) values, consequently providing a criterion for susceptibility testing and clinical usage of enrofloxacin.