EMAIL THIS PAGE TO A FRIEND
Luminescence : the journal of biological and chemical luminescence
Development of a highly sensitive chemiluminescence enzyme immunoassay using enhanced luminol as substrate.
PMID 23785024
Abstract
In this study, a high sensitivity chemiluminescence enzyme immunoassay (CLEIA) based on novel enhancers was developed. Under optimal conditions, we developed an enhanced chemiluminescence reaction (ECR) catalyzed by horseradish peroxidase (HRP-C) in the presence of 3-(10'-phenothiazinyl) propane-1-sulfonate (SPTZ) and 4-morpholinopyridine (MORP) as enhancers. The limit of detection of the newly prepared chemiluminescent cocktail for HRP was 0.33 pg/well, which is lower than that of commercial Super Signal substrate. The results showed that this novel chemiluminescent cocktail can significantly increase the light output of HRP-catalyzed ECR, which can be translated into a corresponding improvement in sensitivity. Similar improvements were observed in CLEIA for the determination of chloramphenicol in milk. In addition, the ECR of N-azoles as secondary enhancer was also presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
480657
4-Morpholinopyridine, 97%
C9H12N2O