EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Reward-modulated motor information in identified striatum neurons.
PMID 23785137
Abstract
It is widely accepted that dorsal striatum neurons participate in either the direct pathway (expressing dopamine D1 receptors) or the indirect pathway (expressing D2 receptors), controlling voluntary movements in an antagonistically balancing manner. The D1- and D2-expressing neurons are activated and inactivated, respectively, by dopamine released from substantia nigra neurons encoding reward expectation. However, little is known about the functional representation of motor information and its reward modulation in individual striatal neurons constituting the two pathways. In this study, we juxtacellularly recorded the spike activity of single neurons in the dorsolateral striatum of rats performing voluntary forelimb movement in a reward-predictable condition. Some of these neurons were identified morphologically by a combination of juxtacellular visualization and in situ hybridization for D1 mRNA. We found that the striatal neurons exhibited distinct functional activations before and during the forelimb movement, regardless of the expression of D1 mRNA. They were often positively, but rarely negatively, modulated by expecting a reward for the correct motor response. The positive reward modulation was independent of behavioral differences in motor performance. In contrast, regular-spiking and fast-spiking neurons in any layers of the motor cortex displayed only minor and unbiased reward modulation of their functional activation in relation to the execution of forelimb movement. Our results suggest that the direct and indirect pathway neurons cooperatively rather than antagonistically contribute to spatiotemporal control of voluntary movements, and that motor information is subcortically integrated with reward information through dopaminergic and other signals in the skeletomotor loop of the basal ganglia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4639
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
C10H16N2O3S
B0301
Biotin, meets USP testing specifications
C10H16N2O3S
B1116000
Biotin, European Pharmacopoeia (EP) Reference Standard
C10H16N2O3S
PHR1233
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
C10H16N2O3S
91827
Biotin, certified reference material, TraceCERT®
C10H16N2O3S
14400
Biotin, ≥99.0% (T)
C10H16N2O3S
19606
Biotin, tested according to Ph.Eur.
C10H16N2O3S
11096176001
Blocking Reagent, For nucleic acid hybridization and detection
00268 Dextran, analytical standard, for GPC, 1,000
00269 Dextran, analytical standard, for GPC, 5,000
00270 Dextran, analytical standard, for GPC, 12,000
00271 Dextran, analytical standard, for GPC, 25,000
00891 Dextran, analytical standard, for GPC, 50,000
00892 Dextran, analytical standard, for GPC, 80,000
00893 Dextran, analytical standard, for GPC, 150,000
00894 Dextran, analytical standard, for GPC, 270,000
00895 Dextran, analytical standard, for GPC, 410,000
00896 Dextran, analytical standard, for GPC, 670,000
31394 Dextran, enzymatic synth.
31430 Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
31416 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
31417 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
D9260 Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
31418 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
D4626 Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
31419 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
D1662 Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
31420 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 50,000
D3759 Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
31421 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 80,000