EMAIL THIS PAGE TO A FRIEND
Voprosy virusologii
[Estimation of activity of bis-netropsin derivatives based on a model of an experimental cutaneous herpes simplex virus disease of guinea pigs].
PMID 23785759
Abstract
Using the model of an experimental cutaneous infection of guinea pig males caused by herpes simple virus type 1, it is shown that application of dimerico derivatives of netropsin Lys-bis Nt and 15Lys-bis Nt in the form of polietilenglicol-based ointment suppresses viral infection more effectively than acyclovir.