EMAIL THIS PAGE TO A FRIEND
Molekuliarnaia genetika, mikrobiologiia i virusologiia
[Development of the high-throughput fluorescence assay detecting SNPs in hemostasis and folate metabolism genes for clinical use].
PMID 23785789
Abstract
Genetic predisposition of an individual patient should be taken in account to choose the proper treatment. Implementation to clinical practice requires the development of rapid, high-throughput, and easy assays intended to detect single nucleotide polymorphisms. A detection kit intended to identify the hemostasis and folate cycle gene mutations G20210A FII, G1691A FV, G10976A FVII, G103T FXIII, C807T ITGA2, T1565C ITGB3, 5G(-675)4G PAI, G(-455)A FGB, C677T and A1298C MTHFR, A2756G MTR, A66G MTRR was suggested in this work. The method is based on the polymerase chain reaction and subsequent melt curve analysis of the complexes of amplicons with specific probe. Three single nucleotide polymorphisms can be identified in one tube using our detection kit that increases the productivity of the analysis in the clinical use. Different types of biological samples (buccal epithelium, saliva, plasma, serum, and urogenital swabs) can be used as the initial material for DNA isolation and further analysis by the method developed in this work.