EMAIL THIS PAGE TO A FRIEND
Journal of the Indian Medical Association
Comparison of results of three different methods of treatment for adhesive capsulitis of shoulder.
PMID 23785922
Abstract
In prospective randomised fashion 72 patients were selected for treatment and were assigned to one of three groups: Groups M, I, and P and treated by manipulation under anaesthesia(M), peri-articular injection (I) and by physiotherapy (P) and results were compared. Though the efficacy of different methods are comparable, but groups M and I have better outcome than group P.