EMAIL THIS PAGE TO A FRIEND
Likars'ka sprava
[I/D angiotensin-converting enzyme gene polymorphism: structural changes in the myocardium and predictive value in chronic heart failure].
PMID 23786008
Abstract
The article presents data regarding links of I/D angiotensin-converting enzyme (ACE) gene polymorphism and structural changes in the myocardium and predictive value in chronic heart failure (CHF). We did not find association of I/D ACE gene polymorphism and structural changes in the myocardium and predictive value in patients with CHF, receiving treatment inhibitor ACE.