EMAIL THIS PAGE TO A FRIEND
Likars'ka sprava
[Clinical and laboratory characteristics of the acetonemia syndrome in children with severe acute intestinal infection].
PMID 23786011
Abstract
A total of 55 children suffering from acute intestinal infection severe in age from 3 months to 7 years; of these, 37 patients with atsetonemicheskim syndrome (AS). Found that the development AS in children with acute intestinal infections severe, aggravate the disease. In children with acute intestinal infection with the syndrome, the duration of atsetonemicheskim main symptoms of intoxication in the 1,2-1,5 times longer than those of children suffering from acute intestinal infection without atsetonemicheskogo syndrome.